missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-iodo-5-nitrobenzene, 98%, Thermo Scientific™

Catalog No. AC378500010
Change view
Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378500010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC378500010 Supplier Thermo Scientific Chemicals Supplier No. 378500010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3819-88-3
Molecular Formula C6H3FINO2
Molecular Weight (g/mol) 266.99
MDL Number MFCD00007219
InChI Key MXPYCSFCKXSPAB-UHFFFAOYSA-N
Synonym 3-fluoro-5-iodonitrobenzene, benzene, 1-fluoro-3-iodo-5-nitro, acmc-1cr1w, 5-fluoro-3-nitroiodobenzene, ksc495i2h, 1-iodo-3-fluoro-5-nitrobenzene, 1-fluoro-3-iodo-5-nitro-benzene, 1-fluoro-3-iodo-5-nitrobenzene
PubChem CID 259086
IUPAC Name 1-fluoro-3-iodo-5-nitrobenzene
SMILES C1=C(C=C(C=C1F)I)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NO2C6H3I(F)
Quantity 1g
Formula Weight 266.99
Percent Purity 98%
Chemical Name or Material 1-Fluoro-3-iodo-5-nitrobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only