accessibility menu, dialog, popup

UserName

NNCl

CAS RN 38180-46-0

IUPAC Name: 3-chloropyridine-2-carbonitrile
Synonyms: 3-chloropyridine-2-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: YDPLFBIGFQFIDB-UHFFFAOYSA-N
SMILES: ClC1=C(N=CC=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results