missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-cyanopyridine 98.0+%, TCI America™

Catalog No. C30225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30225G 5 g
C30221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30225G Supplier TCI America Supplier No. C30225G

Chemical Identifiers

CAS 38180-46-0
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.55
MDL Number MFCD03788834
InChI Key YDPLFBIGFQFIDB-UHFFFAOYSA-N
Synonym 3-chloro-2-cyanopyridine, 2-cyano-3-chloropyridine, 3-chloropicolinonitrile, 3-chloro-2-pyridinecarbonitrile, 2-pyridinecarbonitrile, 3-chloro, 3-chloro-pyridine-2-carbonitrile, 3-chlorpyridin-2-carbonitril, chloro-2-cyanopyridine, pubchem14368, acmc-209iyb
PubChem CID 818258
IUPAC Name 3-chloropyridine-2-carbonitrile
SMILES ClC1=C(N=CC=C1)C#N

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 100°C
Quantity 5 g
UN Number 3439
Formula Weight 138.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-cyanopyridine