CAS RN 381716-33-2
IUPAC Name:
[3,4,5-tris(acetyloxy)-6-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}oxan-2-yl]methyl acetate
Synonyms:
[3,4,5-tris(acetyloxy)-6-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}oxan-2-yl]methyl acetate
Molecular Weight (g/mol):
505.48
Molecular Formula:
C20H31N3O12
InChi Key:
DLTVNCZYHKTWGV-UHFFFAOYNA-N
SMILES:
CC(=O)OCC1OC(OCCOCCOCCN=[N+]=[N-])C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
1
–
1
of
1
results
1
2-[2-(2-Azidoethoxy)ethoxy]ethyl 2,3,4,6-Tetra-O-acetyl-D-galactopyranoside 95.0+%, TCI America™
CAS: 381716-33-2 Molecular Formula: C20H31N3O12 Molecular Weight (g/mol): 505.477 MDL Number: MFCD00191441 InChI Key: DLTVNCZYHKTWGV-IBBQXXDQSA-N PubChem CID: 44630315 IUPAC Name: [(2R,3S,4S,5R)-3,4,5-triacetyloxy-6-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]oxan-2-yl]methyl acetate SMILES: CC(=O)OCC1C(C(C(C(O1)OCCOCCOCCN=[N+]=[N-])OC(=O)C)OC(=O)C)OC(=O)C