missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(2-Azidoethoxy)ethoxy]ethyl 2,3,4,6-Tetra-O-acetyl-D-galactopyranoside 95.0+%, TCI America™

Catalog No. G02575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02575G 5 g
G02571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02575G Supplier TCI America Supplier No. G02575G

Chemical Identifiers

CAS 381716-33-2
Molecular Formula C20H31N3O12
Molecular Weight (g/mol) 505.477
MDL Number MFCD00191441
InChI Key DLTVNCZYHKTWGV-IBBQXXDQSA-N
PubChem CID 44630315
IUPAC Name [(2R,3S,4S,5R)-3,4,5-triacetyloxy-6-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OCCOCCOCCN=[N+]=[N-])OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Quantity 5 g
Formula Weight 505.48
Percent Purity ≥95.0% (N)
Physical Form Liquid
Chemical Name or Material 2-[2-(2-Azidoethoxy)ethoxy]ethyl 2,3,4,6-Tetra-O-acetyl-D-galactopyranoside