accessibility menu, dialog, popup

UserName

CH3OFFF

CAS RN 381-88-4

IUPAC Name: 1,1,1-trifluorobutan-2-one
Synonyms: 1,1,1-trifluorobutan-2-one
Molecular Weight (g/mol): 126.08
Molecular Formula: C4H5F3O
InChi Key: QBVHMPFSDVNFAY-UHFFFAOYSA-N
SMILES: CCC(=O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results