missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-butanone, 96%

Catalog No. AAL1686806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1686806 5 g
AAL1686803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1686806 Supplier Thermo Scientific Chemicals Supplier No. L1686806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 381-88-4 | C4H5F3O | 126.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 381-88-4
Molecular Formula C4H5F3O
Molecular Weight (g/mol) 126.08
MDL Number MFCD00190641
InChI Key QBVHMPFSDVNFAY-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2-butanone, 1,1,1-trifluorobutane-2-one, 2-butanone, 1,1,1-trifluoro, 1,1,1-trifluoromethyl ethyl ketone, trifluoro-2-butanone, acmc-20ap8w, trifluoromethylethylketone, ethyl trifluoromethyl ketone, trifluoromethyl ethyl ketone, 1,1-trifluoro-2-butanone
PubChem CID 238288
IUPAC Name 1,1,1-trifluorobutan-2-one
SMILES CCC(=O)C(F)(F)F

Specifications

Density 0.929
Boiling Point 44°C to 45°C
Flash Point −17°C (1°F)
Refractive Index 1.307
Quantity 5 g
UN Number UN1224
Beilstein 1702220
Formula Weight 126.08
Percent Purity 96%
Chemical Name or Material 1,1,1-Trifluoro-2-butanone
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only