accessibility menu, dialog, popup

UserName

CAS RN 380481-66-3

IUPAC Name: 5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane
Synonyms: 5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane
Molecular Weight (g/mol): 204.08
Molecular Formula: C12H17BO2
InChi Key: ZSPBWRPQSPRISS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)B1OCC(C)(C)CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results