missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzeneboronic acid neopentyl glycol ester, 99%

Catalog No. AAL1956306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1956306 5 g
AAL1956303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1956306 Supplier Thermo Scientific Chemicals Supplier No. L1956306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 380481-66-3 | C12H17BO2 | 204.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380481-66-3
Molecular Formula C12H17BO2
Molecular Weight (g/mol) 204.076
MDL Number MFCD03788728
InChI Key ZSPBWRPQSPRISS-UHFFFAOYSA-N
Synonym 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane
PubChem CID 23005406
IUPAC Name 5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C

Specifications

Melting Point 93°C to 95°C
Quantity 5 g
Formula Weight 204.08
Percent Purity 99%
Chemical Name or Material 4-Methylbenzeneboronic acid neopentyl glycol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only