accessibility menu, dialog, popup

UserName

CAS RN 37908-96-6

IUPAC Name: 3-chloro-4-methoxybenzoate
Synonyms: 3-chloro-4-methoxybenzoate
Molecular Weight (g/mol): 185.58
Molecular Formula: C8H6ClO3
InChi Key: IBCQUQXCTOPJOD-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1Cl)C([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)