accessibility menu, dialog, popup

UserName

CAS RN 3788-94-1

IUPAC Name: ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate
Synonyms: ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: FNASCUBBFNCFQO-VURMDHGXSA-N
SMILES: CCO\C=C(\C(C)=O)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results