missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Acetyl-3-ethoxyacrylate 98.0+%, TCI America™

Catalog No. E11485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11485G 5 g
E114825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11485G Supplier TCI America Supplier No. E11485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3788-94-1
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD06797311
InChI Key FNASCUBBFNCFQO-VURMDHGXSA-N
Synonym 2-Acetyl-3-ethoxyacrylic Acid Ethyl Ester
PubChem CID 5355337
IUPAC Name ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate
SMILES CCOC=C(C(=O)C)C(=O)OCC

Specifications

Boiling Point 266°C
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Acetyl-3-ethoxyacrylate