accessibility menu, dialog, popup

UserName

H3CO

CAS RN 3777-69-3

IUPAC Name: 2-pentylfuran
Synonyms: amylfuran
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: YVBAUDVGOFCUSG-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=CO1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)