missing translation for 'onlineSavingsMsg'
Learn More

2-Pentylfuran 98.0+%, TCI America™

Catalog No. P12095ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12095ML 5 mL
P120925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12095ML Supplier TCI America Supplier No. P12095ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3777-69-3
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00036497
InChI Key YVBAUDVGOFCUSG-UHFFFAOYSA-N
Synonym furan, 2-pentyl, 2-n-pentylfuran, 2-amylfuran, furan, pentyl, pentylfuran, natural, unii-6i0qaj1jzq, 2-pentyl furan, fema no. 3317, 6i0qaj1jzq
PubChem CID 19602
IUPAC Name 2-pentylfuran
SMILES CCCCCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 mL
UN Number 1993
Formula Weight 138.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentylfuran
RTECSNumber LU5187000
TSCA Yes