accessibility menu, dialog, popup

UserName

CH3H3C(S)HNO(S)NNH2H2NNHONH(S)HOOONH(S)NNH2NH2O

CAS RN 37691-11-5

IUPAC Name: (2S)-2-({[(1S)-4-[(diaminomethylidene)amino]-1-{[(1S)-1-{[(2S)-5-[(diaminomethylidene)amino]-1-oxopentan-2-yl]carbamoyl}-2-methylpropyl]carbamoyl}butyl]carbamoyl}amino)-3-phenylpropanoic acid
Synonyms: antipain
Molecular Weight (g/mol): 604.71
Molecular Formula: C27H44N10O6
InChi Key: SDNYTAYICBFYFH-TUFLPTIASA-N
SMILES: CC(C)[C@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O)C(=O)N[C@@H](CCCN=C(N)N)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results