accessibility menu, dialog, popup

UserName

OO

CAS RN 3769-42-4

IUPAC Name: 1,3-bis(benzyloxy)benzene
Synonyms: 1,3-bis(benzyloxy)benzene
Molecular Weight (g/mol): 290.36
Molecular Formula: C20H18O2
InChi Key: RESHZVQZWMQUMB-UHFFFAOYSA-N
SMILES: C(OC1=CC(OCC2=CC=CC=C2)=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results