missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibenzyloxybenzene 96.0+%, TCI America™

Catalog No. B212925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B212925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B212925G Supplier TCI America Supplier No. B212925G
Only null left

Chemical Identifiers

CAS 3769-42-4
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.36
MDL Number MFCD00690156
InChI Key RESHZVQZWMQUMB-UHFFFAOYSA-N
Synonym 1,3-Bis(benzyloxy)benzene, Resorcinol Dibenzyl Ether
PubChem CID 1363616
IUPAC Name 1,3-bis(benzyloxy)benzene
SMILES C(OC1=CC(OCC2=CC=CC=C2)=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 74°C
Quantity 25 g
Formula Weight 290.36
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibenzyloxybenzene