accessibility menu, dialog, popup

UserName

CAS RN 37687-24-4

IUPAC Name: 3,5-diethyl 1H-pyrazole-3,5-dicarboxylate
Synonyms: 3,5-diethyl 1H-pyrazole-3,5-dicarboxylate
Molecular Weight (g/mol): 212.21
Molecular Formula: C9H12N2O4
InChi Key: MBWXLICVQZUJOW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=NN1)C(=O)OCC
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)