missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrazole-3,5-dicarboxylate 98.0+%, TCI America™

Catalog No. D35075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35075G 5 g
D35071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35075G Supplier TCI America Supplier No. D35075G

Chemical Identifiers

CAS 37687-24-4
Molecular Formula C9H12N2O4
Molecular Weight (g/mol) 212.21
MDL Number MFCD00152167
InChI Key MBWXLICVQZUJOW-UHFFFAOYSA-N
Synonym diethyl 3,5-pyrazoledicarboxylate, diethyl pyrazole-3,5-dicarboxylate, 3,5-diethoxycarbonylpyrazole, 1h-pyrazole-3,5-dicarboxylic acid diethyl ester, 3,5-pyrazoledicarboxylic acid diethyl ester, 3,5-diethyl 1h-pyrazole-3,5-dicarboxylate, 1h-pyrazole-3,5-dicarboxylic acid, diethyl ester, zlchem 861, pubchem22955, acmc-1aj76
PubChem CID 142184
IUPAC Name 3,5-diethyl 1H-pyrazole-3,5-dicarboxylate
SMILES CCOC(=O)C1=CC(=NN1)C(=O)OCC

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl Pyrazole-3,5-dicarboxylate