accessibility menu, dialog, popup

UserName

CAS RN 37612-52-5

IUPAC Name: 1-(3-chloro-4-methoxyphenyl)ethan-1-one
Synonyms: 1-(3-chloro-4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: QILWOKAXHOAFOF-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(C=C1)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results