accessibility menu, dialog, popup

UserName

ClONClO

CAS RN 3739-94-4

IUPAC Name: pyridine-2,6-dicarbonyl dichloride
Synonyms: pyridine-2,6-dicarbonyl dichloride
Molecular Weight (g/mol): 204.01
Molecular Formula: C7H3Cl2NO2
InChi Key: GWHOGODUVLQCEB-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(=N1)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)