missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,6-dicarbonyl dichloride, 97%

Catalog No. AAH2657014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2657014 25 g
AAH2657003 1 g
AAH2657006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2657014 Supplier Thermo Scientific Chemicals Supplier No. H2657014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3739-94-4 | C7H3Cl2NO2 | 204.006 g/mol

2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff?s bases. It was used in the synthesis of N,N?-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff′s bases. It was used in the synthesis of N,N′-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 3739-94-4
Molecular Formula C7H3Cl2NO2
Molecular Weight (g/mol) 204.006
MDL Number MFCD00006289
InChI Key GWHOGODUVLQCEB-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153
PubChem CID 77335
IUPAC Name pyridine-2,6-dicarbonyl chloride
SMILES C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl

Specifications

Melting Point 57°C to 61°C
Boiling Point 284°C
Odor Irritating
Quantity 25 g
UN Number UN3261
Beilstein 131556
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 204.01
Percent Purity 98%
Chemical Name or Material Pyridine-2,6-dicarbonyl dichloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 223-125-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only