accessibility menu, dialog, popup

UserName

CAS RN 3724-10-5

IUPAC Name: 2-(methylsulfanyl)benzoic acid
Synonyms: benzoic acid, o-(methylthio)-
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: LWJQGKJCZOGGPJ-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)