missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzoic Acid 98.0+%, TCI America™

Catalog No. M19535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19535G 5 g
M195325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19535G Supplier TCI America Supplier No. M19535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3724-10-5
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00029934
InChI Key LWJQGKJCZOGGPJ-UHFFFAOYSA-N
Synonym 2-methylthio benzoic acid, benzoic acid, 2-methylthio, o-methylthio benzoic acid, benzoic acid, o-methylthio, 2-methylsulfanyl benzoic acid, 2-methylmercaptobenzoic acid, 2-carboxyphenyl methyl sulfide, acide methyl-s-2-benzoique, 2-methylmercapto benzoic acid, acide methyl-s-2-benzoique french
PubChem CID 19498
IUPAC Name 2-methylsulfanylbenzoic acid
SMILES CSC1=CC=CC=C1C(=O)O

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 168.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)benzoic Acid
RTECSNumber DH4390000
TSCA No