accessibility menu, dialog, popup

UserName

CAS RN 370865-89-7

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; [(cyanoimino)methylidene]azanide
Synonyms: 1-ethyl-3-methylimidazolium; [(cyanoimino)methylidene]azanide
Molecular Weight (g/mol): 177.21
Molecular Formula: C8H11N5
InChi Key: MKHFCTXNDRMIDR-UHFFFAOYSA-N
SMILES: [N-]=C=NC#N.CCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)