missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium dicyanamide, 98%

Catalog No. AAH2690118
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2690118 50 g
AAH2690106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2690118 Supplier Thermo Scientific Chemicals Supplier No. H2690118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 370865-89-7 | C8H11N5 | 177.211 g/mol

Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Soluble in water. Soluble in solvents like methanol, acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 370865-89-7
Molecular Formula C8H11N5
Molecular Weight (g/mol) 177.211
MDL Number MFCD08276373
InChI Key MKHFCTXNDRMIDR-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium dicyanamide, ethylmethylimidazolium dicyanamide, emim dca, acmc-209iqn, basionics™ vs 03, dsstox_cid_27940, dsstox_rid_82692, dsstox_gsid_47964, ksc221g0h
PubChem CID 11159638
IUPAC Name cyanoiminomethylideneazanide;1-ethyl-3-methylimidazol-3-ium
SMILES CCN1C=C[N+](=C1)C.C(=[N-])=NC#N

Specifications

Quantity 50 g
Solubility Information Soluble in water. Soluble in solvents like methanol,acetone.
Formula Weight 177.21
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium dicyanamide
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only