CAS RN 3682-32-4
IUPAC Name:
4-hydroxy-3-nitrosonaphthalene-1-sulfonic acid
Synonyms:
4-hydroxy-3-nitrosonaphthalene-1-sulfonic acid
Molecular Weight (g/mol):
253.23
Molecular Formula:
C10H7NO5S
InChi Key:
GASPSNIEUBWWJU-UHFFFAOYSA-N
SMILES:
OC1=C2C=CC=CC2=C(C=C1N=O)S(O)(=O)=O
1
–
1
of
1
results
1
4-Hydroxy-3-nitroso-1-naphthalenesulfonic Acid Hydrate 98.0+%, TCI America™
CAS: 3682-32-4 Molecular Formula: C10H7NO5S Molecular Weight (g/mol): 253.228 MDL Number: MFCD00003933 InChI Key: GASPSNIEUBWWJU-UHFFFAOYSA-N Synonym: 2-nitroso-1-naphthol-4-sulfonic acid,4-hydroxy-3-nitroso-1-naphthalenesulfonic acid,1-naphthalenesulfonic acid, 4-hydroxy-3-nitroso,unii-7t06pca43o,4-hydroxy-3-nitroso-naphthalene-1-sulfonic acid,4-hydroxy-3-nitrosonaphthalenesulfonic acid,nitroso-nw acid,acmc-1cril,4-hydroxy-3-nitrosonaphthalene-1-sulphonic acid,1-naphthalenesulfonicacid, 4-hydroxy-3-nitroso PubChem CID: 77257 IUPAC Name: 4-hydroxy-3-nitrosonaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2O)N=O)S(=O)(=O)O