accessibility menu, dialog, popup

UserName

CAS RN 36519-61-6

IUPAC Name: 10-methyl-9-phenylacridin-10-ium perchlorate
Synonyms: 10-methyl-9-phenylacridin-10-ium perchlorate
Molecular Weight (g/mol): 369.8
Molecular Formula: C20H16ClNO4
InChi Key: NRNCXGCGIKATGE-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.C[N+]1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results