accessibility menu, dialog, popup

UserName

CAS RN 3641-51-8

CH3(R)HOOOHO

CAS RN 3641-51-8

IUPAC Name: (2R)-2-methylbutanedioic acid
Synonyms: (2R)-2-methylbutanedioic acid
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: WXUAQHNMJWJLTG-GSVOUGTGSA-N
SMILES: C[C@H](CC(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)