missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-Methylsuccinic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 114°C |
| Color | Purple-White |
| Quantity | 5 g |
| Formula Weight | 132.12 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-(+)-Methylsuccinic Acid |
Safety and Handling
| EINECSNumber | (2)-0903 |
| TSCA | No |