CAS RN 3627-01-8
IUPAC Name:
4-hydroxy-5-{2-[(1E)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonic acid
Synonyms:
4-hydroxy-5-{2-[(1E)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonic acid
Molecular Weight (g/mol):
440.4
Molecular Formula:
C16H12N2O9S2
InChi Key:
CYJIGBWVVZHXRF-SFQUDFHCSA-N
SMILES:
OC1=CC(=O)C=C\C1=N/NC1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
1
–
1
of
1
results
1
H-Resorcinol 97.0+%, TCI America™
CAS: 3627-01-8 Molecular Formula: C16H12N2O9S2 Molecular Weight (g/mol): 440.40 MDL Number: MFCD00059024 InChI Key: CYJIGBWVVZHXRF-SFQUDFHCSA-N Synonym: 1-(2,4-Dihydroxy-1-phenylazo)-8-hydroxynaphthalene-3,6-disulfonic Acid Disodium Salt PubChem CID: 9575828 IUPAC Name: 4-hydroxy-5-[(2E)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid SMILES: C1=CC(=NNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=CC1=O)O