accessibility menu, dialog, popup

UserName

CAS RN 362-43-6

IUPAC Name: 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 284.27
Molecular Formula: C12H16N2O6
InChi Key: GFDUSNQQMOENLR-PEBGCTIMSA-N
SMILES: CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=CC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results