missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Isopropylideneuridine

Catalog No. AAL0368614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0368614 25 g
AAL0368603 1 g
AAL0368606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0368614 Supplier Thermo Scientific Chemicals Supplier No. L0368614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 362-43-6 | C12H16N2O6 | 284.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 362-43-6
Molecular Formula C12H16N2O6
Molecular Weight (g/mol) 284.268
MDL Number MFCD00034509
InChI Key GFDUSNQQMOENLR-PEBGCTIMSA-N
Synonym 2',3'-isopropylideneuridine, 2',3'-o-isopropylidene uridine, 2',3'-o-isopropylideneuridine, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4 1h,3h-dione, 1-1r,2r,4r,5r-4-hydroxymethyl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oct-2-yl-1,3-dihydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-3h-pyrimidine-2,4-dione, 2/',3/'-isopropylideneuridine, 2',3'-o-isopropylideneuridine hplc, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 64967
IUPAC Name 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione
SMILES CC1(OC2C(OC(C2O1)N3C=CC(=O)NC3=O)CO)C

Specifications

Melting Point 165°C to 166°C
Quantity 25 g
Beilstein 32671
Formula Weight 284.27
Chemical Name or Material 2',3'-Isopropylideneuridine
EINECSNumber 206-647-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only