CAS RN 36176-31-5
IUPAC Name:
1-isothiocyanato-2-(propan-2-yl)benzene
Synonyms:
1-isopropyl-2-isothiocyanatobenzene
Molecular Weight (g/mol):
177.27
Molecular Formula:
C10H11NS
InChi Key:
QQOOMDSEMQHMIR-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC=C1N=C=S
1
–
1
of
1
results
1
Promotions Available
2-Isopropylphenyl isothiocyanate, 96%
2-Isopropylphenyl isothiocyanate, 96%
CAS: 36176-31-5 Molecular Formula: C10H11NS Molecular Weight (g/mol): 177.265 MDL Number: MFCD00022053 InChI Key: QQOOMDSEMQHMIR-UHFFFAOYSA-N Synonym: 2-isopropylphenyl isothiocyanate,1-isopropyl-2-isothiocyanatobenzene,1-isothiocyanato-2-propan-2-yl benzene,benzene, 1-isothiocyanato-2-1-methylethyl,2-methylethyl benzenisothiocyanate,2-isopropylphenylisothiocyanate,acmc-20ansf,2-isopropylphenyl-isothiocyanate,2-iso-propylphenyl isothiocyanate PubChem CID: 142052 IUPAC Name: 1-isothiocyanato-2-propan-2-ylbenzene SMILES: CC(C)C1=CC=CC=C1N=C=S