missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl isothiocyanate, 96%

Catalog No. AAA1944814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1944814 25 g
AAA1944806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1944814 Supplier Thermo Scientific Chemicals Supplier No. A1944814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36176-31-5 | C10H11NS | 177.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36176-31-5
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.265
MDL Number MFCD00022053
InChI Key QQOOMDSEMQHMIR-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isothiocyanate, 1-isopropyl-2-isothiocyanatobenzene, 1-isothiocyanato-2-propan-2-yl benzene, benzene, 1-isothiocyanato-2-1-methylethyl, 2-methylethyl benzenisothiocyanate, 2-isopropylphenylisothiocyanate, acmc-20ansf, 2-isopropylphenyl-isothiocyanate, 2-iso-propylphenyl isothiocyanate
PubChem CID 142052
IUPAC Name 1-isothiocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=S

Specifications

Density 1.05
Boiling Point 131°C to 132°C (10 mmHg)
Refractive Index 1.605
Quantity 25 g
UN Number UN2922
Beilstein 2717112
Sensitivity Moisture sensitive
Formula Weight 177.27
Percent Purity 96%
Chemical Name or Material 2-Isopropylphenyl isothiocyanate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only