accessibility menu, dialog, popup

UserName

CAS RN 3615-56-3

IUPAC Name: (3R,4S,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: d-sorbose D-xylo-hex-2-ulose D-xylo-hexulose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: LKDRXBCSQODPBY-IANNHFEVSA-N
SMILES: OCC1(O)OC[C@@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)