missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Sorbose, 98%

Catalog No. AAB2120877
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2120877 0.1 g
AAB2120802 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2120877 Supplier Thermo Scientific Chemicals Supplier No. B2120877

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3615-56-3 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3615-56-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00151095
InChI Key LKDRXBCSQODPBY-IANNHFEVSA-N
Synonym d-sorbose, keto-d-sorbose, unii-jj09461njs, 3r,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, xylo-hexulose, sorbose, d, d-sor, d-xylo-hex-2-ulose, d-+-sorbose
PubChem CID 107428
ChEBI CHEBI:13022
SMILES OCC1(O)OC[C@@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 163°C to 165°C
Quantity 0.1 g
IUPAC Name (3R,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material D-Sorbose
EINECSNumber 222-796-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only