accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNNH2

CAS RN 361345-40-6

IUPAC Name: tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 291.4
Molecular Formula: C16H25N3O2
InChi Key: VKSINZNBWVJILC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC2=CC(N)=CC=C2)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)