accessibility menu, dialog, popup

UserName

NN

CAS RN 357645-40-0

IUPAC Name: N2,N7-bis(naphthalen-1-yl)-N2,N7,9,9-tetraphenyl-9H-fluorene-2,7-diamine
Synonyms: N2,N7-bis(naphthalen-1-yl)-N2,N7,9,9-tetraphenylfluorene-2,7-diamine
Molecular Weight (g/mol): 752.96
Molecular Formula: C57H40N2
InChi Key: PQCAURRJHOJJNQ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC2=C(C=C1)C1=C(C=C(C=C1)N(C1=CC=CC=C1)C1=C3C=CC=CC3=CC=C1)C2(C1=CC=CC=C1)C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results