missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(1-naphthyl)-N,N',9,9-tetraphenyl-9H-fluorene-2,7-diamine 98.0+%, TCI America™

Catalog No. D5443200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5443200MG 200 mg
D54431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5443200MG Supplier TCI America Supplier No. D5443200MG

Chemical Identifiers

CAS 357645-40-0
Molecular Formula C57H40N2
Molecular Weight (g/mol) 752.96
MDL Number MFCD11112145
InChI Key PQCAURRJHOJJNQ-UHFFFAOYSA-N
Synonym DPFL-NPB, 9,9-Diphenyl-2,7-bis[N-(1-naphthyl)anilino]fluorene
PubChem CID 53384285
IUPAC Name N2,N7-bis(naphthalen-1-yl)-N2,N7,9,9-tetraphenyl-9H-fluorene-2,7-diamine
SMILES C1=CC=C(C=C1)N(C1=CC2=C(C=C1)C1=C(C=C(C=C1)N(C1=CC=CC=C1)C1=C3C=CC=CC3=CC=C1)C2(C1=CC=CC=C1)C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 239°C
Quantity 200 mg
Formula Weight 752.96
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di(1-naphthyl)-N,N′,9,9-tetraphenyl-9H-fluorene-2,7-diamine