accessibility menu, dialog, popup

UserName

CAS RN 35762-87-9

NNSOOF

CAS RN 35762-87-9

IUPAC Name: pyrimidine-2-sulfonyl fluoride
Synonyms: pyrimidine-2-sulfonyl fluoride
Molecular Weight (g/mol): 162.14
Molecular Formula: C4H3FN2O2S
InChi Key: HMZIGHBQOOGZAK-UHFFFAOYSA-N
SMILES: FS(=O)(=O)C1=NC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results