accessibility menu, dialog, popup

UserName

CAS RN 356-42-3

IUPAC Name: 2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
Synonyms: 2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
Molecular Weight (g/mol): 310.05
Molecular Formula: C6F10O3
InChi Key: XETRHNFRKCNWAJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)