accessibility menu, dialog, popup

UserName

CAS RN 35512-87-9

BH3CPCH3

CAS RN 35512-87-9

IUPAC Name: dimethyl(phenyl)phosphane boron
Synonyms: dimethylphenylphosphine boron
Molecular Weight (g/mol): 148.96
Molecular Formula: C8H11BP
InChi Key: QZAXONBUUURRQX-UHFFFAOYSA-N
SMILES: [B].CP(C)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results