missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylphenylphosphine Borane 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 78°C |
| Quantity | 5 g |
| Formula Weight | 151.98 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dimethylphenylphosphine Borane |
Safety and Handling
| TSCA | No |