accessibility menu, dialog, popup

UserName

CAS RN 355-34-0

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexan-1-amine
Synonyms: 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexan-1-amine
Molecular Weight (g/mol): 299.09
Molecular Formula: C6H4F11N
InChi Key: FDIHRNYGDZQEAV-UHFFFAOYSA-N
SMILES: NCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results