missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Undecafluorohexylamine 97.0+%, TCI America™

Catalog No. U00835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00835G 5 g
U00831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00835G Supplier TCI America Supplier No. U00835G

Chemical Identifiers

CAS 355-34-0
Molecular Formula C6H4F11N
Molecular Weight (g/mol) 299.087
MDL Number MFCD09025929
InChI Key FDIHRNYGDZQEAV-UHFFFAOYSA-N
Synonym 1H,1H-Perfluorohexylamine
PubChem CID 21923864
IUPAC Name 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexan-1-amine
SMILES C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)N

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 g
UN Number 2735
Formula Weight 299.09
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1H,1H-Undecafluorohexylamine