accessibility menu, dialog, popup

UserName

CAS RN 354-38-1

IUPAC Name: trifluoroacetamide
Synonyms: trifluoroacetamide
Molecular Weight (g/mol): 113.04
Molecular Formula: C2H2F3NO
InChi Key: NRKYWOKHZRQRJR-UHFFFAOYSA-N
SMILES: NC(=O)C(F)(F)F
Boiling Point 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (5)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)