missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetamide, 97%

Catalog No. AAA1445136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1445136 500 g
AAA1445114 25 g
AAA1445122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1445136 Supplier Thermo Scientific Chemicals Supplier No. A1445136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 354-38-1 | C2H2F3NO | 113.04 g/mol

2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.

Solubility
Soluble in water (460 g/L at 20°C), alcohol, ether, chloroform (partially), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.

Chemical Identifiers

CAS 354-38-1
Molecular Formula C2H2F3NO
Molecular Weight (g/mol) 113.04
MDL Number MFCD00008008
InChI Key NRKYWOKHZRQRJR-UHFFFAOYSA-N
Synonym trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622
PubChem CID 67717
IUPAC Name 2,2,2-trifluoroacetamide
SMILES NC(=O)C(F)(F)F

Specifications

Melting Point 70°C to 75°C
Boiling Point 162°C to 164°C
Quantity 500 g
Beilstein 1753625
Solubility Information Soluble in water (460g/L at 20°C),alcohol,ether,chloroform (partially),and methanol.
Formula Weight 113.04
Percent Purity 97%
Chemical Name or Material 2,2,2-Trifluoroacetamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-559-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only