accessibility menu, dialog, popup

UserName

CH3NHOCH3OCH3

CAS RN 34956-31-5

IUPAC Name: N-(4-acetyl-3-methylphenyl)acetamide
Synonyms: N-(4-acetyl-3-methylphenyl)acetamide
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: PTARWPNYVATTDE-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(C)=C(C=C1)C(C)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)