missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetamido-2'-methylacetophenone 98.0+%, TCI America™

Catalog No. A27675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27675G 5 g
A276725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27675G Supplier TCI America Supplier No. A27675G

Chemical Identifiers

CAS 34956-31-5
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD02258876
InChI Key PTARWPNYVATTDE-UHFFFAOYSA-N
Synonym n-4-acetyl-3-methylphenyl acetamide, 4'-acetamido-2'-methylacetophenone, 4-acetamido-2-methylacetophenone, pubchem12562, 4'-acetyl-3'-methylacetoanilide, n-4-acetyl-3-methyl-phenyl-acetamide, 4\\'-acetamido-2\\'-methylacetophenone, n-4-ethanoyl-3-methyl-phenyl ethanamide, acetamide, n-4-acetyl-3-methylphenyl
PubChem CID 2737381
IUPAC Name N-(4-acetyl-3-methylphenyl)acetamide
SMILES CC(=O)NC1=CC(C)=C(C=C1)C(C)=O

Specifications

Melting Point 139°C
Color Yellow
Quantity 5 g
Formula Weight 191.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Acetamido-2′-methylacetophenone